accessibility menu, dialog, popup

UserName

CAS RN 6138-79-0

HH2OClCH3NN(E)

CAS RN 6138-79-0

IUPAC Name: hydrogen 2-[(1E)-1-(4-methylphenyl)-3-(pyrrolidin-1-yl)prop-1-en-1-yl]pyridine hydrate chloride
Synonyms: hydrogen triprolidine hydrate chloride
Molecular Weight (g/mol): 332.87
Molecular Formula: C19H25ClN2O
InChi Key: CUZMOIXUFHOLLN-UMVVUDSKSA-N
SMILES: [H+].O.[Cl-].CC1=CC=C(C=C1)C(=C/CN1CCCC1)\C1=CC=CC=N1
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results