accessibility menu, dialog, popup

UserName

CAS RN 61341-26-2

IUPAC Name: 3-methylthiophene-2-carbonyl chloride
Synonyms: 3-methylthiophene-2-carbonyl chloride
Molecular Weight (g/mol): 160.62
Molecular Formula: C6H5ClOS
InChi Key: AJSVPEVDFBYRCH-UHFFFAOYSA-N
SMILES: CC1=C(SC=C1)C(Cl)=O
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)