accessibility menu, dialog, popup

UserName

FINH2

CAS RN 61272-76-2

IUPAC Name: 4-fluoro-2-iodoaniline
Synonyms: 4-fluoro-2-iodoaniline
Molecular Weight (g/mol): 237.02
Molecular Formula: C6H5FIN
InChi Key: SETOTRGVPANENO-UHFFFAOYSA-N
SMILES: NC1=C(I)C=C(F)C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)