missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-iodoaniline, 96%

Catalog No. AAB2549914
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2549914 25 g
AAB2549906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2549914 Supplier Thermo Scientific Chemicals Supplier No. B2549914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 61272-76-2 | C6H5FIN | 237.02 g/mol

4-Fluoro-2-iodoaniline is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluoro-2-iodoaniline is used as an organic chemical synthesis intermediate.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 61272-76-2
Molecular Formula C6H5FIN
Molecular Weight (g/mol) 237.02
MDL Number MFCD00042584
InChI Key SETOTRGVPANENO-UHFFFAOYSA-N
Synonym 2-iodo-4-fluoroaniline, 4-fluoro-2-indoaniline, 4-fluoro-2-iodoanline, 4-fluoro-2-iodo-aniline, 4-fluoro-2-iodophenylamine, 4-fluoro-2-iodo-phenylamine, benzenamine, 4-fluoro-2-iodo, zlchem 368, pubchem3413, 2-iodo-4-fluoro-aniline
PubChem CID 2778283
IUPAC Name 4-fluoro-2-iodoaniline
SMILES NC1=C(I)C=C(F)C=C1

Specifications

Quantity 25 g
UN Number UN2810
Solubility Information Not miscible in water.
Formula Weight 237.02
Percent Purity 98%
Chemical Name or Material 4-Fluoro-2-iodoaniline
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only