missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-iodoaniline 98.0+%, TCI America™

Catalog No. F07065G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F07065G 5 g
F070625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F07065G Supplier TCI America Supplier No. F07065G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 61272-76-2
Molecular Formula C6H5FIN
Molecular Weight (g/mol) 237.02
MDL Number MFCD00042584
InChI Key SETOTRGVPANENO-UHFFFAOYSA-N
Synonym 2-iodo-4-fluoroaniline, 4-fluoro-2-indoaniline, 4-fluoro-2-iodoanline, 4-fluoro-2-iodo-aniline, 4-fluoro-2-iodophenylamine, 4-fluoro-2-iodo-phenylamine, benzenamine, 4-fluoro-2-iodo, zlchem 368, pubchem3413, 2-iodo-4-fluoro-aniline
PubChem CID 2778283
IUPAC Name 4-fluoro-2-iodoaniline
SMILES NC1=C(I)C=C(F)C=C1

Specifications

Quantity 5 g
Formula Weight 237.02
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Fluoro-2-iodoaniline