accessibility menu, dialog, popup

UserName

CAS RN 61135-33-9

IUPAC Name: 5-ethynyl-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms: 5-ethynyl-2'-deoxyuridine
Molecular Weight (g/mol): 252.23
Molecular Formula: C11H12N2O5
InChi Key: CDEURGJCGCHYFH-DJLDLDEBSA-N
SMILES: OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(C#C)C(=O)NC1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results