missing translation for 'onlineSavingsMsg'
Learn More

5-Ethynyl-2'-deoxyuridine 98.0+%, TCI America™

Catalog No. E105750MG
Change view
Click to view available options
Quantity:
50 mg
200 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E105750MG 50 mg
E1057200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E105750MG Supplier TCI America Supplier No. E105750MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 61135-33-9
Molecular Formula C11H12N2O5
Molecular Weight (g/mol) 252.23
MDL Number MFCD01675687
InChI Key CDEURGJCGCHYFH-DJLDLDEBSA-N
Synonym EdU
PubChem CID 472172
IUPAC Name 5-ethynyl-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione
SMILES OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(C#C)C(=O)NC1=O

Specifications

Melting Point 199°C
Color White-Yellow
Quantity 50 mg
Formula Weight 252.23
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Ethynyl-2′-deoxyuridine
RTECSNumber YU7510000
TSCA No