accessibility menu, dialog, popup

UserName

FClO

CAS RN 61072-56-8

IUPAC Name: 4-chloro-2-fluorobenzaldehyde
Synonyms: 4-chloro-2-fluorobenzaldehyde
Molecular Weight (g/mol): 158.56
Molecular Formula: C7H4ClFO
InChi Key: UVGYSEIWAOOIJR-UHFFFAOYSA-N
SMILES: FC1=CC(Cl)=CC=C1C=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)