missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-fluorobenzaldehyde, 98%

Catalog No. AAA1611009
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1611009 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1611009 Supplier Thermo Scientific Chemicals Supplier No. A1611009
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 61072-56-8 | C7H4ClFO | 158.556 g/mol

4-Chloro-2-fluorobenzaldehyde is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chloro-2-fluorobenzaldehyde is used as pharmaceutical intermediates.

Solubility
Insoluble in water.

Notes
Air sensitive. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 61072-56-8
Molecular Formula C7H4ClFO
Molecular Weight (g/mol) 158.556
MDL Number MFCD00143282
InChI Key UVGYSEIWAOOIJR-UHFFFAOYSA-N
Synonym 2-fluoro-4-chlorobenzaldehyde, benzaldehyde, 4-chloro-2-fluoro, 4-chloranyl-2-fluoranyl-benzaldehyde, pubchem1435, acmc-1b5ai, ksc494c2f, 3-fluoro-4-formylchlorobenzene, 4-chloro-2-fluoro-benzaldehyde, 4-chloro-2-fluoro-benzoaldehyde, timtec-bb sbb003985
PubChem CID 2724908
IUPAC Name 4-chloro-2-fluorobenzaldehyde
SMILES C1=CC(=C(C=C1Cl)F)C=O

Specifications

Melting Point 58°C to 60°C
Quantity 10 g
Beilstein 3537705
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 158.56
Percent Purity 98%
Chemical Name or Material 4-Chloro-2-fluorobenzaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only