missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-fluorobenzaldehyde 98.0+%, TCI America™

Catalog No. C15975G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C15975G 5 g
C159725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C15975G Supplier TCI America Supplier No. C15975G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 61072-56-8
Molecular Formula C7H4ClFO
Molecular Weight (g/mol) 158.556
MDL Number MFCD00143282
InChI Key UVGYSEIWAOOIJR-UHFFFAOYSA-N
Synonym 2-fluoro-4-chlorobenzaldehyde, benzaldehyde, 4-chloro-2-fluoro, 4-chloranyl-2-fluoranyl-benzaldehyde, pubchem1435, acmc-1b5ai, ksc494c2f, 3-fluoro-4-formylchlorobenzene, 4-chloro-2-fluoro-benzaldehyde, 4-chloro-2-fluoro-benzoaldehyde, timtec-bb sbb003985
PubChem CID 2724908
IUPAC Name 4-chloro-2-fluorobenzaldehyde
SMILES C1=CC(=C(C=C1Cl)F)C=O

Specifications

Melting Point 60°C
Color White-Yellow
Quantity 5 g
Formula Weight 158.56
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-2-fluorobenzaldehyde
TSCA No
Recommended Storage Refrigerator