accessibility menu, dialog, popup

UserName

CAS RN 610-81-1

IUPAC Name: 4-amino-3-nitrophenol
Synonyms: 4-amino-3-nitrophenol
Molecular Weight (g/mol): 154.13
Molecular Formula: C6H6N2O3
InChi Key: IQXUIDYRTHQTET-UHFFFAOYSA-N
SMILES: NC1=CC=C(O)C=C1[N+]([O-])=O
Molecular Weight (g/mol) 
  • (4)
  • (3)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (2)
  • (3)
  • (2)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (4)
  • (3)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (5)