missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-nitrophenol, 98%

Catalog No. AAL0948714
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0948714 25 g
AAL0948706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0948714 Supplier Thermo Scientific Chemicals Supplier No. L0948714
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 610-81-1 | C6H6N2O3 | 154.125 g/mol

4-Amino-3-nitrophenol was used in the synthesis of benzimidazole based Factor Xa (trypsin-like serine protease) inhibitors and 4-(4-amino-3-nitrophenoxy)phthalonitrile. It was also employed as matrix during visible matrix-assisted laser desorption/ionization mass spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Amino-3-nitrophenol was used in the synthesis of benzimidazole based Factor Xa (trypsin-like serine protease) inhibitors and 4-(4-amino-3-nitrophenoxy)phthalonitrile. It was also employed as matrix during visible matrix-assisted laser desorption/ionization mass spectrometry.

Solubility
Soluble in water. (1.8 g/L.)

Notes
Store at room temperature. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 610-81-1
Molecular Formula C6H6N2O3
Molecular Weight (g/mol) 154.125
MDL Number MFCD00066310
InChI Key IQXUIDYRTHQTET-UHFFFAOYSA-N
Synonym 4-hydroxy-2-nitroaniline, 3-nitro-4-aminophenol, phenol, 4-amino-3-nitro, 4-amino-3-nitro-phenol, unii-r5wy41q95z, 3-nitro-4-amino phenol, 4-aminophenol-3-nitrophenol, pubchem1481, 4-amino-3-nitropbenol
PubChem CID 3758882
IUPAC Name 4-amino-3-nitrophenol
SMILES C1=CC(=C(C=C1O)[N+](=O)[O-])N

Specifications

Melting Point 148°C to 153°C
Odor Characteristic
Quantity 25 g
Beilstein 2210196
Solubility Information Soluble in water. (1.8g/L.)
Formula Weight 154.13
Percent Purity 98%
Chemical Name or Material 4-Amino-3-nitrophenol
Hazard Category H302-H315-H319-H335-H341
Hazard Statement GHS H Statement
H302-H312-H319
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c
EINECSNumber 210-236-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only