accessibility menu, dialog, popup

UserName

CAS RN 609-65-4

IUPAC Name: 2-chlorobenzoyl chloride
Synonyms: 2-chlorobenzoyl chloride
Molecular Weight (g/mol): 175.01
Molecular Formula: C7H4Cl2O
InChi Key: ONIKNECPXCLUHT-UHFFFAOYSA-N
SMILES: ClC(=O)C1=CC=CC=C1Cl
Boiling Point 
  • (3)
  • (3)
  • (1)
Percent Purity 
  • (3)
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (4)
  • (3)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)