accessibility menu, dialog, popup

UserName

CAS RN 609-25-6

IUPAC Name: 5-methylbenzene-1,2,3-triol
Synonyms: 5-methylbenzene-1,2,3-triol
Molecular Weight (g/mol): 140.14
Molecular Formula: C7H8O3
InChi Key: NYUABOGYMWADSF-UHFFFAOYSA-N
SMILES: CC1=CC(O)=C(O)C(O)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results