accessibility menu, dialog, popup

UserName

CAS RN 60835-74-7

IUPAC Name: 2,3,5,6,8,9,11,12,14,15-decahydro-1,4,7,10,13,16-benzohexaoxacyclooctadecine-18-carbaldehyde
Synonyms: 2,3,5,6,8,9,11,12,14,15-decahydro-1,4,7,10,13,16-benzohexaoxacyclooctadecine-18-carbaldehyde
Molecular Weight (g/mol): 340.37
Molecular Formula: C17H24O7
InChi Key: ALMXRNQJWRGNMG-UHFFFAOYSA-N
SMILES: O=CC1=CC2=C(OCCOCCOCCOCCOCCO2)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results