accessibility menu, dialog, popup

UserName

CAS RN 608-43-5

IUPAC Name: 2,3-dimethylbenzene-1,4-diol
Synonyms: 1,4-benzenediol, 2,3-dimethyl-
Molecular Weight (g/mol): 138.17
Molecular Formula: C8H10O2
InChi Key: BXJGUBZTZWCMEX-UHFFFAOYSA-N
SMILES: CC1=C(C)C(O)=CC=C1O
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)