missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethylhydroquinone, 97%

Catalog No. AC305111000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC305111000 100 g Glass bottle
AC305110050 5 g Glass bottle
AC305110250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC305111000 Supplier Thermo Scientific Chemicals Supplier No. 305111000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 608-43-5 | C8H10O2 | 138.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 608-43-5
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.17
MDL Number MFCD00009997
InChI Key BXJGUBZTZWCMEX-UHFFFAOYSA-N
Synonym 2,3-dimethylhydroquinone, o-xylene-3,6-diol, 2,3-xylohydroquinone, o-xylohydroquinone, 1,4-benzenediol, 2,3-dimethyl, unii-f0l7hg609j, 1,4-dihydroxy-2,3-dimethylbenzene, 3,6-dihydroxy-o-xylene, 2,3-dimethyl-1,4-benzenediol, 2,3-dimethyl hydroquinone
PubChem CID 69100
IUPAC Name 2,3-dimethylbenzene-1,4-diol
SMILES CC1=C(C)C(O)=CC=C1O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 223.0°C to 225.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 100 g
Solubility Information Solubility in water: slightly soluble in water
Formula Weight 138.17
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 2, 3-Dimethylhydroquinone
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER

RUO – Research Use Only