accessibility menu, dialog, popup

UserName

CH3H3CI

CAS RN 608-28-6

IUPAC Name: 2-iodo-1,3-dimethylbenzene
Synonyms: benzene, 2-iodo-1,3-dimethyl-
Molecular Weight (g/mol): 232.06
Molecular Formula: C8H9I
InChi Key: QTUGGVBKWIYQSS-UHFFFAOYSA-N
SMILES: CC1=CC=CC(C)=C1I
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)