accessibility menu, dialog, popup

UserName

HOO

CAS RN 607-42-1

IUPAC Name: anthracene-1-carboxylic acid
Synonyms: 1-anthracenecarboxylic acid
Molecular Weight (g/mol): 222.24
Molecular Formula: C15H10O2
InChi Key: CCFAKBRKTKVJPO-UHFFFAOYSA-N
SMILES: OC(=O)C1=C2C=C3C=CC=CC3=CC2=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results