missing translation for 'onlineSavingsMsg'
Learn More

1-Anthracenecarboxylic Acid 99.0+%, TCI America™

Catalog No. A12201G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A12201G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A12201G Supplier TCI America Supplier No. A12201G
Only null left

Chemical Identifiers

CAS 607-42-1
Molecular Formula C15H10O2
Molecular Weight (g/mol) 222.24
MDL Number MFCD00142583
InChI Key CCFAKBRKTKVJPO-UHFFFAOYSA-N
PubChem CID 150789
ChEBI CHEBI:38994
IUPAC Name anthracene-1-carboxylic acid
SMILES OC(=O)C1=C2C=C3C=CC=CC3=CC2=CC=C1

Specifications

Melting Point 252°C
Quantity 1 g
Formula Weight 222.24
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Anthracenecarboxylic Acid