accessibility menu, dialog, popup

UserName

FOFFF

CAS RN 60611-24-7

IUPAC Name: 2-fluoro-6-(trifluoromethyl)benzaldehyde
Synonyms: 2-fluoro-6-(trifluoromethyl)benzaldehyde
Molecular Weight (g/mol): 192.11
Molecular Formula: C8H4F4O
InChi Key: FAKUGVHRTLCKHB-UHFFFAOYSA-N
SMILES: FC1=CC=CC(=C1C=O)C(F)(F)F
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)