missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Fluoro-6-(trifluoromethyl)benzaldehyde, 98%, Thermo Scientific™
Specifications
Specifications
| Boiling Point | 156°C |
| Flash Point | 82°C |
| Assay Percent Range | 97.5% min. (GC) |
| Packaging | Glass Bottle |
| Quantity | 1g |
| Formula Weight | 192.11 |
| Percent Purity | 98% |
| Chemical Name or Material | 2-Fluoro-6-(trifluoromethyl)benzaldehyde |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
RUO – Research Use Only