accessibility menu, dialog, popup

UserName

CH3OOOCH3

CAS RN 606-45-1

IUPAC Name: methyl 2-methoxybenzoate
Synonyms: methyl O-anisate
Molecular Weight (g/mol): 166.18
Molecular Formula: C9H10O3
InChi Key: PFYHAAAQPNMZHO-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CC=C1OC
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)