accessibility menu, dialog, popup

UserName

CAS RN 60491-10-3

IUPAC Name: bis((11S,18S,20R,21R,22S)-12-oxa-8,17-diazaheptacyclo[15.5.2.0¹,¹⁸.0²,⁷.0⁸,²².0¹¹,²¹.0¹⁵,²⁰]tetracosa-2,4,6,14-tetraen-9-one) sulfuric acid pentahydrate
Synonyms: bis((11S,18S,20R,21R,22S)-12-oxa-8,17-diazaheptacyclo[15.5.2.0¹,¹⁸.0²,⁷.0⁸,²².0¹¹,²¹.0¹⁵,²⁰]tetracosa-2,4,6,14-tetraen-9-one) sulfuric acid pentahydrate
Molecular Weight (g/mol): 856.99
Molecular Formula: C42H56N4O13S
InChi Key: FDQUTDUZMOOVST-YSVQHLMNSA-N
SMILES: O.O.O.O.O.OS(O)(=O)=O.O=C1C[C@@H]2OCC=C3CN4CCC56[C@@H]4C[C@@H]3[C@@H]2[C@@H]5N1C1=CC=CC=C61.O=C1C[C@@H]2OCC=C3CN4CCC56[C@@H]4C[C@@H]3[C@@H]2[C@@H]5N1C1=CC=CC=C61
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results