accessibility menu, dialog, popup

UserName

H2OCuCH3OOCH3OO

CAS RN 6046-93-1

IUPAC Name: copper(2+) diacetate hydrate
Synonyms: copper(2+) diacetate hydrate
Molecular Weight (g/mol): 199.65
Molecular Formula: C4H8CuO5
InChi Key: NWFNSTOSIVLCJA-UHFFFAOYSA-L
SMILES: O.[Cu++].CC([O-])=O.CC([O-])=O
Quantity 
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (6)
  • (1)
Physical Form 
  • (5)
  • (8)
  • (3)
  • (3)
Percent Purity 
  • (2)
  • (6)
  • (5)
  • (3)
  • (2)
Grade 
  • (3)
  • (3)
  • (5)
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (8)
  • (8)
  • (2)
  • (14)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (6)
  • (1)
  • (5)
  • (8)
  • (3)
  • (3)
  • (2)
  • (6)
  • (5)
  • (3)
  • (2)
  • (3)
  • (3)
  • (5)
  • (3)
  • (3)