missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) Acetate Monohydrate 95.0+%, TCI America™

Catalog No. C2346500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2346500G 500 g
C234625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C2346500G Supplier TCI America Supplier No. C2346500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6046-93-1
Molecular Formula C4H8CuO5
Molecular Weight (g/mol) 199.65
MDL Number MFCD00149570
InChI Key NWFNSTOSIVLCJA-UHFFFAOYSA-L
Synonym copper ii acetate monohydrate, cupric acetate monohydrate, copper diacetate monohydrate, copper acetate monohydrate, copper 2+ acetate, monohydrate, acetic acid, copper 2+ salt, monohydrate, verdigris, copper acetate hydrate, diacetoxycopper hydrate, copper acetate, hydrate
PubChem CID 165397
IUPAC Name copper(2+) diacetate hydrate
SMILES O.[Cu++].CC([O-])=O.CC([O-])=O

Specifications

Melting Point 115°C
Color Blue
pH 5.2 to 5.5 (20g/L 20°C)
Quantity 500 g
UN Number 3077
Formula Weight 181.63
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Copper(II) Acetate Monohydrate
EINECSNumber (2)-0693
RTECSNumber AG3500000
TSCA Yes