missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) acetate monohydrate, 99.9% (metals basis)

Catalog No. AA3548114
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3548114 25 g
AA3548106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3548114 Supplier Thermo Scientific Chemicals Supplier No. 03548114

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6046-93-1 | C4H8CuO5 | 199.65 g/mol

Copper(II) acetate monohydrate is used in formation of Cu chelates of β-dicarbonyl compounds as a means of purification and in the eglinton (modified glaser) coupling of terminal alkynes. It is also used as a catalyst for the michael reaction giving increased yields and absence of side-reactions. It Promotes ullmann-type C-O and C-N coupling reactions of arylboronic acids with phenols, amines and various other nitrogen derivatives. It is also used in biochemical applications such as DNA extraction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6046-93-1
Molecular Formula C4H8CuO5
Molecular Weight (g/mol) 199.65
MDL Number MFCD00149570
InChI Key NWFNSTOSIVLCJA-UHFFFAOYSA-L
Synonym copper ii acetate monohydrate, cupric acetate monohydrate, copper diacetate monohydrate, copper acetate monohydrate, copper 2+ acetate, monohydrate, acetic acid, copper 2+ salt, monohydrate, verdigris, copper acetate hydrate, diacetoxycopper hydrate, copper acetate, hydrate
PubChem CID 165397
IUPAC Name copper;diacetate;hydrate
SMILES O.[Cu++].CC([O-])=O.CC([O-])=O

Specifications

Melting Point 115°C
Color Blue-Green
pH 5.2 to 5.5
Quantity 25 g
Odor Odorless
Density 1.882 g/mL
Linear Formula Cu(OOCCH3)2·H2O
UN Number UN3077
Merck Index 14,2624
Solubility Information Soluble in water (72mg/ml at 20°C),alcohol,ether (slightly),and glycerin (slightly).
Formula Weight 199.65 (181.64 Anhydrous)
Percent Purity 99.9%
Assay (metals basis)
Chemical Name or Material Copper(II) acetate monohydrate
Show More Show Less
Hazard Category H302-H314-H335-H373
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 205-553-3
RTECSNumber AG3500000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only