missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper(II) acetate monohydrate, ACS reagent
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
Specifications
Specifications
| Boiling Point | 240.0°C |
| Melting Point | 115.0°C |
| Color | Blue-Green or Green |
| Physical Form | Solid |
| Quantity | 500 g |
| Infrared Spectrum | Authentic |
| Packaging | Plastic bottle |
| Density | 1.88g/mL |
| Linear Formula | (CH3CO2)2Cu·H2O |
| Fieser | 01,159; 02,84; 03,65; 04,105; 05,156; 06,138; 10,103; 12,140; 14,199; 15,99; 17,176 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Harmful if swallowed. Causes severe skin burns and eye damage. Very toxic to aquatic life with long lasting effects. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with wat |
RUO – Research Use Only