accessibility menu, dialog, popup

UserName

CAS RN 60421-23-0

HClCH3OOH2N

CAS RN 60421-23-0

IUPAC Name: hydrogen methyl 1-aminocyclopentane-1-carboxylate chloride
Synonyms: hydrogen methyl 1-aminocyclopentane-1-carboxylate chloride
Molecular Weight (g/mol): 179.64
Molecular Formula: C7H14ClNO2
InChi Key: OPUJUITUYWGUEP-UHFFFAOYSA-N
SMILES: [H+].[Cl-].COC(=O)C1(N)CCCC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results