CAS RN 6036-48-2
IUPAC Name:
naphthalene-1-sulfonic acid
Synonyms:
α-naphthalenesulfonic acid
Molecular Weight (g/mol):
208.23
Molecular Formula:
C10H8O3S
InChi Key:
PSZYNBSKGUBXEH-UHFFFAOYSA-N
SMILES:
OS(=O)(=O)C1=C2C=CC=CC2=CC=C1
1
–
1
of
1
results
1
Promotions Available
Naphthalene-1-sulfonic acid hydrate, 98%
Naphthalene-1-sulfonic acid hydrate, 98%
CAS: 6036-48-2 Molecular Formula: C10H8O3S Molecular Weight (g/mol): 208.23 MDL Number: MFCD00003987 InChI Key: PSZYNBSKGUBXEH-UHFFFAOYSA-N Synonym: naphthalene-1-sulfonic acid dihydrate,1-naphthalenesulfonic acid dihydrate,naphthalene-1-sulfonic acid hydrate PubChem CID: 71431272 IUPAC Name: naphthalene-1-sulfonic acid;dihydrate SMILES: OS(=O)(=O)C1=C2C=CC=CC2=CC=C1