accessibility menu, dialog, popup

UserName

CAS RN 60301-20-4

H3CNNH2N

CAS RN 60301-20-4

IUPAC Name: 1-methyl-1H-indazol-3-amine
Synonyms: 1-methylindazol-3-amine
Molecular Weight (g/mol): 147.18
Molecular Formula: C8H9N3
InChi Key: NYLGITXFVVEBLZ-UHFFFAOYSA-N
SMILES: CN1N=C(N)C2=CC=CC=C12
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results