accessibility menu, dialog, popup

UserName

CAS RN 602-55-1

IUPAC Name: 9-phenylanthracene
Synonyms: 9-phenylanthracene
Molecular Weight (g/mol): 254.33
Molecular Formula: C20H14
InChi Key: LUBXLGUQZVKOFP-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)C1=C2C=CC=CC2=CC2=CC=CC=C12
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results