accessibility menu, dialog, popup

UserName

H2NHNNHClHCl

CAS RN 5993-91-9

IUPAC Name: 1-(1H-1,3-benzodiazol-2-yl)methanamine dihydrochloride
Synonyms: 1-(1H-1,3-benzodiazol-2-yl)methanamine dihydrochloride
Molecular Weight (g/mol): 220.1
Molecular Formula: C8H11Cl2N3
InChi Key: HAEYZZSSSUIZAN-UHFFFAOYSA-N
SMILES: Cl.Cl.NCC1=NC2=CC=CC=C2N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results