accessibility menu, dialog, popup

UserName

CH3BrF

CAS RN 59907-12-9

IUPAC Name: 1-bromo-2-fluoro-3-methylbenzene
Synonyms: 1-bromo-2-fluoro-3-methylbenzene
Molecular Weight (g/mol): 189.03
Molecular Formula: C7H6BrF
InChi Key: LZVNGSFAHGKCDM-UHFFFAOYSA-N
SMILES: CC1=CC=CC(Br)=C1F
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)