missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-fluorotoluene, 98%, Thermo Scientific™

Catalog No. AAH6149214
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6149214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6149214 Supplier Thermo Scientific Chemicals Supplier No. H6149214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 59907-12-9
Molecular Formula C7H6BrF
Molecular Weight (g/mol) 189.027
MDL Number MFCD04972694
InChI Key LZVNGSFAHGKCDM-UHFFFAOYSA-N
Synonym 3-bromo-2-fluorotoluene, 1-bromo-2-fluoro-3-methyl-benzene, benzene, 1-bromo-2-fluoro-3-methyl, 2-fluoro-3-bromo toluene, 2-fluoro-3-methyl-bromobenzene, pubchem4521, a-bromo-a-fluorotoluene, 2-fluoro-3-bromotoluene, acmc-1b1zk, ksc493o4d
PubChem CID 108855
IUPAC Name 1-bromo-2-fluoro-3-methylbenzene
SMILES CC1=C(C(=CC=C1)Br)F

Specifications

Melting Point 118°C to 123°C
Density 1.503 g/mL
Boiling Point 118°C to 123°C
Flash Point 76°C (169°F)
Refractive Index 1.533
Quantity 25 g
Formula Weight 189.02
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Bromo-2-fluorotoluene
Hazard Category H227-H302-H315-H319-H335
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
EINECSNumber 261-981-0
TSCA No
Recommended Storage Ambient temperatures