missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-fluorotoluene 98.0+%, TCI America™

Catalog No. B29695G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B29695G 5 g
B29691G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B29695G Supplier TCI America Supplier No. B29695G

Chemical Identifiers

CAS 59907-12-9
Molecular Formula C7H6BrF
Molecular Weight (g/mol) 189.027
MDL Number MFCD04972694
InChI Key LZVNGSFAHGKCDM-UHFFFAOYSA-N
Synonym 3-bromo-2-fluorotoluene, 1-bromo-2-fluoro-3-methyl-benzene, benzene, 1-bromo-2-fluoro-3-methyl, 2-fluoro-3-bromo toluene, 2-fluoro-3-methyl-bromobenzene, pubchem4521, a-bromo-a-fluorotoluene, 2-fluoro-3-bromotoluene, acmc-1b1zk, ksc493o4d
PubChem CID 108855
IUPAC Name 1-bromo-2-fluoro-3-methylbenzene
SMILES CC1=C(C(=CC=C1)Br)F

Specifications

Color Yellow
Boiling Point 186°C
Quantity 5 g
Formula Weight 189.03
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Bromo-2-fluorotoluene