accessibility menu, dialog, popup

UserName

CAS RN 599-61-1

IUPAC Name: 3-(3-aminobenzenesulfonyl)aniline
Synonyms: benzenamine, 3,3'-sulfonylbis-
Molecular Weight (g/mol): 248.3
Molecular Formula: C12H12N2O2S
InChi Key: LJGHYPLBDBRCRZ-UHFFFAOYSA-N
SMILES: NC1=CC(=CC=C1)S(=O)(=O)C1=CC(N)=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)