accessibility menu, dialog, popup

UserName

CAS RN 59803-98-4

IUPAC Name: 5-bromo-N-(4,5-dihydro-1H-imidazol-2-yl)quinoxalin-6-amine
Synonyms: brimonidine brimonidine tartrate alphagan bromoxidine allergan brand of brimonidine tartrate brimonidine (product) brimonidine purite brimonidina ratio-brimonidine brimonidine preparation
Molecular Weight (g/mol): 292.14
Molecular Formula: C11H10BrN5
InChi Key: XYLJNLCSTIOKRM-UHFFFAOYSA-N
SMILES: BrC1=C2N=CC=NC2=CC=C1NC1=NCCN1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)