accessibility menu, dialog, popup

UserName

CAS RN 598-16-3

BrBrBr

CAS RN 598-16-3

IUPAC Name: 1,1,2-tribromoethene
Synonyms: ethene, tribromo-
Molecular Weight (g/mol): 264.74
Molecular Formula: C2HBr3
InChi Key: OZVJKTHTULCNHB-UHFFFAOYSA-N
SMILES: BrC=C(Br)Br
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results