accessibility menu, dialog, popup

UserName

CH3OH3C

CAS RN 5973-71-7

IUPAC Name: 3,4-dimethylbenzaldehyde
Synonyms: 3,4-dimethylbenzaldehyde
Molecular Weight (g/mol): 134.18
Molecular Formula: C9H10O
InChi Key: POQJHLBMLVTHAU-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=O)C=C1C
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (6)
  • (4)
  • (2)
  • (2)
  • (4)
  • (3)
  • (3)
  • (4)