missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethylbenzaldehyde, 97%

Catalog No. AAA1538814
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1538814 25 g
AAA1538806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1538814 Supplier Thermo Scientific Chemicals Supplier No. A1538814
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 5973-71-7 | C9H10O | 134.178 g/mol

3,4-Dimethylbenzaldehyde is used as an additive for resins, agricultural intermediate and for flavor and fragrance, nucleation transparent agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dimethylbenzaldehyde is used as an additive for resins, agricultural intermediate and for flavor and fragrance, nucleation transparent agent.

Solubility
Soluble in ethanol, ether, toluene. Insoluble in water.

Notes
Air sensitive. Store under inert gas. Keep way from oxidizing agents, air.

Chemical Identifiers

CAS 5973-71-7
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.178
MDL Number MFCD00016612
InChI Key POQJHLBMLVTHAU-UHFFFAOYSA-N
Synonym benzaldehyde, 3,4-dimethyl, unii-16g1j12arb, 4-formyl-o-xylene, o-xylene-4-carboxaldehyde, 3,4-dimethyl-benzaldehyde, 3 pound not4-dimethylbenzaldehyde, pubchem2535, 3,4 dimethyl benzaldehyde, 3,4-di-methylbenzaldehyde, 3,4-dimethyl benzaldehyde
PubChem CID 22278
IUPAC Name 3,4-dimethylbenzaldehyde
SMILES CC1=C(C=C(C=C1)C=O)C

Specifications

Density 1.017
Boiling Point 226°C
Odor Almond-like
Refractive Index 1.551
Quantity 25 g
Beilstein 1634270
Sensitivity Air Sensitive
Solubility Information Soluble in ethanol,ether,toluene. Insoluble in water.
Formula Weight 134.18
Percent Purity 97%
Chemical Name or Material 3,4-Dimethylbenzaldehyde
Show More Show Less
EINECSNumber 227-770-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only