missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethylbenzaldehyde 95.0+%, TCI America™

Catalog No. D33345G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D33345G 5 g
D333425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D33345G Supplier TCI America Supplier No. D33345G

Chemical Identifiers

CAS 5973-71-7
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.178
MDL Number MFCD00016612
InChI Key POQJHLBMLVTHAU-UHFFFAOYSA-N
Synonym benzaldehyde, 3,4-dimethyl, unii-16g1j12arb, 4-formyl-o-xylene, o-xylene-4-carboxaldehyde, 3,4-dimethyl-benzaldehyde, 3 pound not4-dimethylbenzaldehyde, pubchem2535, 3,4 dimethyl benzaldehyde, 3,4-di-methylbenzaldehyde, 3,4-dimethyl benzaldehyde
PubChem CID 22278
IUPAC Name 3,4-dimethylbenzaldehyde
SMILES CC1=C(C=C(C=C1)C=O)C

Specifications

Color Yellow
Boiling Point 226°C
Quantity 5 g
Formula Weight 134.18
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,4-Dimethylbenzaldehyde
EINECSNumber (3)-3741
TSCA Yes