accessibility menu, dialog, popup

UserName

H3CNHOCH3

CAS RN 5961-59-1

IUPAC Name: 4-methoxy-N-methylaniline
Synonyms: 4-methoxy-N-methylaniline
Molecular Weight (g/mol): 137.18
Molecular Formula: C8H11NO
InChi Key: JFXDIXYFXDOZIT-UHFFFAOYSA-N
SMILES: CNC1=CC=C(OC)C=C1
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)