accessibility menu, dialog, popup

UserName

NH2HNOBr

CAS RN 5933-32-4

IUPAC Name: 4-bromobenzohydrazide
Synonyms: benzoylhydrazine, P-bromo
Molecular Weight (g/mol): 215.05
Molecular Formula: C7H7BrN2O
InChi Key: UYIMBYKIIMYFPS-UHFFFAOYSA-N
SMILES: NNC(=O)C1=CC=C(Br)C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)