missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzhydrazide, 98+%

Catalog No. AAA1891814
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1891814 25 g
AAA1891806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1891814 Supplier Thermo Scientific Chemicals Supplier No. A1891814
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 5933-32-4 | C7H7BrN2O | 215.05 g/mol

4-Bromobenzoic hydrazide has been used in the preparation of {2-[1-oxo-3-(4-bromophenyl)-2-propenyl]hydrazide}-4-bromobenzoic acid, {2,2?-[1,4-phenylenebis(1-oxo-2-propene-3,1-diyl)]dihydrazide}-p-bromobenzoic acid and substituted period[1,2-a]pyrimidine-2,4(3H)-diones (PPMDO).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromobenzoic hydrazide has been used in the preparation of {2-[1-oxo-3-(4-bromophenyl)-2-propenyl]hydrazide}-4-bromobenzoic acid, {2,2′-[1,4-phenylenebis(1-oxo-2-propene-3,1-diyl)]dihydrazide}-p-bromobenzoic acid and substituted period[1,2-a]pyrimidine-2,4(3H)-diones (PPMDO).

Solubility
Soluble in hot methanol (almost transparency). Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 5933-32-4
Molecular Formula C7H7BrN2O
Molecular Weight (g/mol) 215.05
MDL Number MFCD00007602
InChI Key UYIMBYKIIMYFPS-UHFFFAOYSA-N
Synonym 4-bromobenzhydrazide, p-bromobenzhydrazide, 4-bromobenzoic hydrazide, p-bromobenzohydrazide, p-bromobenzoic acid hydrazide, 4-bromobenzoylhydrazine, 4-bromobenzoic acid hydrazide, benzoic acid, 4-bromo-, hydrazide, 4-bromo-benzoic acid hydrazide, p-bromobenzoylhydrazine
PubChem CID 22219
IUPAC Name 4-bromobenzohydrazide
SMILES C1=CC(=CC=C1C(=O)NN)Br

Specifications

Melting Point 167°C to 169°C
Quantity 25 g
Beilstein 777006
Solubility Information Soluble in hot methanol (almost transparency). Slightly soluble in water.
Formula Weight 215.05
Percent Purity ≥98%
Chemical Name or Material 4-Bromobenzhydrazide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 227-681-9
RTECSNumber DG4449250
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only