missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzohydrazide 97,0+%, TCI America™

Numéro de catalogue. B20675G
Modifier l'affichage
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
B20675G 5 g
B206725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. B20675G Fournisseur TCI America Code fournisseur B20675G

Identifiants chimiques

CAS 5933-32-4
Molecular Formula C7H7BrN2O
Molecular Weight (g/mol) 215.05
MDL Number MFCD00007602
InChI Key UYIMBYKIIMYFPS-UHFFFAOYSA-N
Synonym 4-bromobenzhydrazide, p-bromobenzhydrazide, 4-bromobenzoic hydrazide, p-bromobenzohydrazide, p-bromobenzoic acid hydrazide, 4-bromobenzoylhydrazine, 4-bromobenzoic acid hydrazide, benzoic acid, 4-bromo-, hydrazide, 4-bromo-benzoic acid hydrazide, p-bromobenzoylhydrazine
PubChem CID 22219
IUPAC Name 4-bromobenzohydrazide
SMILES C1=CC(=CC=C1C(=O)NN)Br

Spécifications

Melting Point 168°C
Color Blanc
Quantity 5 g
Formula Weight 215.05
Percent Purity ≥97.0% (T)
Physical Form Poudre cristalline
Chemical Name or Material 4-Bromobenzohydrazide
RTECSNumber DG4449250
TSCA No