accessibility menu, dialog, popup

UserName

OONHNOO

CAS RN 5930-93-8

IUPAC Name: 4-nitro-1H-pyrrole-2-carboxylate
Synonyms: 4-nitro-1H-pyrrole-2-carboxylate
Molecular Weight (g/mol): 155.09
Molecular Formula: C5H3N2O4
InChi Key: SNWGRRCTCGASCN-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC(=CN1)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)