accessibility menu, dialog, popup

UserName

CAS RN 593-08-8

IUPAC Name: tridecan-2-one
Synonyms: 2-tridecanone
Molecular Weight (g/mol): 198.35
Molecular Formula: C13H26O
InChi Key: CYIFVRUOHKNECG-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCC(C)=O
Boiling Point 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)