missing translation for 'onlineSavingsMsg'
Learn More

2-Tridecanone, 98+%

Catalog No. AAA1904222
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1904222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1904222 Supplier Thermo Scientific Chemicals Supplier No. A1904222
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 593-08-8 | C13H26O | 198.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 593-08-8
Molecular Formula C13H26O
Molecular Weight (g/mol) 198.35
MDL Number MFCD00008968
InChI Key CYIFVRUOHKNECG-UHFFFAOYSA-N
Synonym 2-tridecanone, methyl undecyl ketone, mathyl undecyl kepoje, hendecyl methyl ketone, 2-tridecankje, tridecanone-2, methyl n-undecyl ketone, tridecanone, 2-tridecanone natural, unii-5q35vhx26k
PubChem CID 11622
ChEBI CHEBI:77928
IUPAC Name tridecan-2-one
SMILES CCCCCCCCCCCC(=O)C

Specifications

Melting Point 29°C to 31°C
Density 0.822
Boiling Point 133°C to 134°C (10 mmHg)
Flash Point 122°C (251°F)
Odor Characteristic
Refractive Index 1.435
Quantity 100 g
UN Number UN3082
Beilstein 1757402
Formula Weight 198.35
Percent Purity ≥98%
Chemical Name or Material 2-Tridecanone
Show More Show Less
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 209-784-0
RTECSNumber YD4574000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only