accessibility menu, dialog, popup

UserName

CAS RN 589-57-1

IUPAC Name: diethyl chlorophosphonite
Synonyms: ethyl phosphorochloridite
Molecular Weight (g/mol): 156.55
Molecular Formula: C4H10ClO2P
InChi Key: TXHWYSOQHNMOOU-UHFFFAOYSA-N
SMILES: CCOP(Cl)OCC
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)